Would you like to log in again?
Factor Xa Activity Fluorometric Assay Kit , BioVision Inc.
Supplier: BioVision
AB204711-100TEST
76022-198EA 934.6 USD InStock
76022-198
Factor Xa Activity Fluorometric Assay Kit , BioVision Inc.
Assays Enzyme Assays
Detect the activity of Factor Xa in plasma
  • Rapid, and convenient
  • The assay can detect Factor Xa activity as low as 1 ng.
Factor Xa (FXa) is the activated form of the coagulation factor X (Stuart-Power factor, thrombokinase, prothrombinase, thromboplastin, E.C.3.4.21.6). Factor X, a serine endopeptidase plays an important role at several stages of the coagulation pathway. It acts by converting prothrombin into active thrombin by complexing with activated co-factor V in the prothrombinase complex. Unfractionated heparin and various low molecular weight heparins bind to plasma cofactor antithrombin to inactivate several coagulation factors including factor Xa. Biovision's Factor Xa activity assay kit utilizes the ability of Factor Xa to cleave a synthetic substrate thereby releasing a fluorophore, AMC, which can be quantified by fluorescence readers. This assay kit is simple, rapid and can detect Factor Xa activity as low as 1 ng.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty