Would you like to log in again?
Succinate Dehydrogenase Activity Colorimetric Assay Kit, BioVision
Supplier: BioVision
AB228560-100TEST
76022-200EA 1191.96 USD InStock
76022-200
Succinate Dehydrogenase Activity Colorimetric Assay Kit, BioVision
Assays Enzyme Assays
Sensitive assay, HTS
  • Simple & rapid
  • High-throughput adaptable
  • This assay kit can detect less than 0.1mU Succinate Dehydrogenase Activity in a variety of samples.
Succinate Dehydrogenase (SDH) (EC 1.3.5.1) or succinate-coenzyme Q reductase (SQR) or respiratory complex II is an enzyme complex, which is bound to the inner mitochondrial membrane. SDH participates in both the citric acid cycle and electron transport chain. In mammals and many bacteria, SDH consists of 2 hydrophilic subunits, SDHA (flavoprotein) and SDHB (iron-sulfur protein) and 2 hydrophobic membrane anchor subunits: SDHC and SDHD. SDH oxidizes succinate to fumarate and transfers the electrons to ubiquinone. SDH deficiency in humans leads to a variety of phenotypes including Leigh syndrome, a neurometabolic disorder, tumor formation, and myopathy. Recent studies show that SDH can prevent the generation of ROS (reactive oxygen species); therefore, measurement of succinate dehydrogenase activity has wide applications. BioVision’s Succinate Dehydrogenase Activity Assay kit is rapid, simple and high-throughput adaptable. In this assay, Succinate dehydrogenase converts succinate to fumarate, and transfers the electron to an artificial electron acceptor (Probe), which changes the color from blue to a colorless product (depending upon the sample enzymatic activity). This assay kit can detect less than 0.1mU Succinate Dehydrogenase Activity in a variety of samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty