Would you like to log in again?
Triose Phosphate Isomerase (TPI) Activity Colorimetric Assay Kit, BioVision
Supplier: BioVision
AB197001-100TEST
76022-206EA 1032.16 USD InStock
76022-206
Triose Phosphate Isomerase (TPI) Activity Colorimetric Assay Kit, BioVision
Assays Enzyme Assays
Only Kit on the market to measure Triose Phosphate Isomerase (TPI) Activity
  • Simple, highly sensitive, and high-throughput.
  • The assay can measure Triose Phosphate Isomerase activity as low as 40 mU/ml.
Triose Phosphate Isomerase (TPI or TIM: EC 5.3.1.1) is an important enzyme for glycolysis. It reversibly interconverts dihydroxyacetone phosphate and glyceraldehyde-3-phosphate, thus maintaining the equilibrium of these two triose phosphates. TPI connects glycolysis to pentose phosphate pathway and lipid metabolism. It is a stable homodimer found in almost all organisms. In humans, TPI deficiency is a rare multisystem disorder and leads to progressive neurological dysfunction, characterized by hemolytic anemia, cardiomyopathy and progressive neuromuscular impairment. BioVision’s Triose Phosphate Isomerase Activity Assay kit provides a quick and easy way for monitoring Triose Phosphate Isomerase activity in a variety of samples. In this kit, Triose Phosphate Isomerase converts dihydroxyacetone phosphate into glyceradehyde-3-phosphate, which reacts with the Enzyme Mix & Developer to form a colored product with strong absorbance at 450 nm. The assay is simple, sensitive, & high-throughput and can detect Triose Phosphate Isomerase activity as low as 40 mU/ml.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty