Would you like to log in again?
Succinyl-CoA Synthetase Activity Colorimetric Assay Kit, BioVision
Supplier: BioVision
AB196989-100TEST
76022-210EA 1381.66 USD InStock
76022-210
Succinyl-CoA Synthetase Activity Colorimetric Assay Kit, BioVision
Assays Enzyme Assays
Only kit on the market to measure Succinyl CoA Synthatase Activity
  • Simple & Rapid Protocol
  • High-Throughput adaptable
  • This kit can detect less than 0.1 mU of Succinyl-CoA Synthetase activity in a variety of samples.
Succinyl-CoA Synthetase (SCS, also called Succinyl-CoA ligase, Succinate Thiokinase) (EC 6.2.1.5) is a critical enzyme in the citric acid cycle and an important metabolic intermediate for porphyrin, heme and ketone body biosynthesis. It is located in the mitochondrial matrix and is a heterodimer composed of one α and one β subunit. In humans, Succinyl-CoA Synthetase deficiency causes the build-up of lactic acid leading to lactic acidosis, which can be fatal in infants. Measurement and analysis of SCS activity is useful for both mechanistic studies as well as for diagnostic purposes. In BioVision’s Succinyl-CoA Synthetase Activity Assay, SCS converts succinate into succinyl-CoA in the presence of ATP and CoA. Succinyl-CoA reacts with the Developer to form a colored product with strong absorbance at 450 nm. This assay kit is simple, sensitive, and high-throughput adaptable. It can detect less than 0.1 mU of Succinyl-CoA Synthetase activity in a variety of samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty