Would you like to log in again?
Fructose Colorimetric/Fluorometric Assay Kit, BioVision Inc.
Supplier: BioVision
AB83380-100
76022-214EA 1011.55 USD InStock
76022-214
Fructose Colorimetric/Fluorometric Assay Kit, BioVision Inc.
Assays Cellular Assays
Sensitive Assay, HTS
  • Simple procedure; takes ~ 1 hour
  • Fast and convenient
  • The sensitive kit contains all necessary reagents for accurate measurement of Fructose
Fructose is a monosaccharide found in many foods and is one of the three most important blood sugars along with glucose and galactose. Fructose is the sweetest naturally occurring sugar, estimated to be twice as sweet as sucrose. In BioVision’s Fructose Assay Kit, free fructose is enzymatically converted to β-glucose, which is then specifically converted to a product that reacts with OxiRed Probe to generate color (λ=570nm) and fluorescence (Ex/Em=535/587nm). The kit provides a rapid, simple, sensitive, and reliable method suitable for high throughput assayof D-fructose.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty