Would you like to log in again?
Invertase Activity Colorimetric Assay Kit, BioVision
Supplier: BioVision
AB197005-100TEST
76022-216EA 814.14 USD InStock
76022-216
Invertase Activity Colorimetric Assay Kit, BioVision
Assays Enzyme Assays
Sensitive Assay, HTS
  • Simple, Sensitive & Rapid protocol
  • Highly-Sensitive: Detects as low as 20 µU Invertase in various sample types
Invertase (EC 3.2.1.26), also known as sucrase or β -fructofuranosidase, catalyzes the hydrolysis of sucrose (table sugar) by cleaving its glycosidic bond and forming one molecule each of glucose and fructose. Invertase is widely expressed among plants and microorganisms including yeast and bacteria. It is extensively utilized in industrial processes, including food, ethanol production, cosmetics, drugs, paper and bioelectronics electrodes. BioVision’s Invertase Activity Assay kit provides a simple, sensitive and high-throughput adaptable assay that detects relevant concentrations of Invertase in biological samples or purified/crude enzyme preparations. The assay is based on the enzymatic detection of glucose, one of the hydrolysis products. Detection limit: 20 µU.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty