Would you like to log in again?
D-Gluconate (D-Gluconic Acid) Assay Kit (Colorimetric), BioVision
Supplier: BioVision
AB204703-100TEST
76022-220EA 1013.06 USD InStock
76022-220
D-Gluconate (D-Gluconic Acid) Assay Kit (Colorimetric), BioVision
Assays Enzyme Assays
Highly sensitive assay to measure D-Gluconic acid
  • Non-radioactive
D-Gluconic acid (C6H12O7) is a mild organic acid that is produced from glucose by glucose oxidase. It is abundantly present in plants, fruits and animal tissues. D-Gluconate (C6H11O7) is the salt or ester of Gluconic Acid. Due to its low toxicity, it is widely used in pharmaceutical, food, and other industries. BioVision’s D-Gluconate (Gluconic Acid) Assay kit is a sensitive, fast and easy-to-use kit. In this assay, Gluconate is utilized by Gluconokinase to form D-Gluconate-6-P and ADP, which subsequently undergoes a series of reactions to form an intermediate that reduces Gluconate probe to give a product with strong absorbance at 450 nm. This assay kit can detect D-Gluconate (D-Gluconic Acid) level less than 2 µM in a variety of samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty