Would you like to log in again?
Cytochrome P450 Reductase (CPR) Activity Kit (Colorimetric), BioVision
Supplier: BioVision
AB204704-100TEST
76022-222EA 1049.24 USD InStock
76022-222
Cytochrome P450 Reductase (CPR) Activity Kit (Colorimetric), BioVision
Assays Enzyme Assays
Unique detection method using a synthetic probe and NADPH generating system.
  • Only 96-well plate format CPR assay kit on the market Only kit to use a synthetic colorimetric probe and a NADPH generating system
  • Rapid, convenient and sensitive
  • Kit contains all necessary reagents for accurate measurement of CPR activity down to 0.2 mU per well in a wide variety of sample types
NADPH-cytochrome P450 reductase (CPR, EC 1.6.2.4) is a ~78 kDa membrane-bound flavoenzyme that catalyzes the transfer of electrons from NADPH to members of the cytochrome P450 monooxidase (CYP) enzyme family in the endoplasmic reticulum. CPR contains two tightly bound flavin cofactors, FAD and FMN, which participate in the sequential transfer of electrons from NADPH→FAD→FMN→CYP, oxidizing NADPH to NADP+ and reducing the CYP heme moiety to the substrate- and oxygen-binding ferrous state. As CPR is required for the function of all CYP isozymes, it plays a critical role in the metabolism of drugs, organic pollutants and other xenobiotic compounds, in addition to its role in biosynthesis of certain vitamins and steroid hormones. BioVision’s cytochrome P450 reductase activity assay kit couples oxidation of NADPH by CPR to reduction of a nearly colorless probe into a brightly colored product with an absorbance peak at 460 nm, with the rate of color generation being directly proportional to CPR activity. The NADPH utilized by CPR is generated in situ from β-NADP+ via oxidation of glucose-6-phosphate (G6P) to 6-phospho-D-glucono-1,5-lactone by glucose-6-phosphatase dehydrogenase (G6PDH). The kit can be used to determine CPR activity in a variety of samples, with a detection limit of ~0.2 mU of CPR activity per reaction. For assessment of CPR activity in crude biological samples that may have extraneous reductases capable of reducing the substrate, an inhibitor of NADPH-dependent flavoproteins is included. In this case, the specific CPR activity may be calculated by running parallel reactions in the presence and absence of inhibitor and subtracting any residual activity detected with the inhibitor present. The kit contains sufficient reagents for performing 100 reactions in a 96-well plate format.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty