Would you like to log in again?
PicoProbe™ Lactate Dehydrogenase Activity Assay Kit , BioVision
Supplier: BioVision
AB197000-500TEST
76022-266EA 957.61 USD InStock
76022-266
PicoProbe™ Lactate Dehydrogenase Activity Assay Kit , BioVision
Assays Enzyme Assays
The most sensitive Kit on the market
  • Simple & highly sensitive
  • High-throughput adaptable
  • Detection Limit: 1 µU/ml
Lactate Dehydrogenase (LDH: EC 1.1.1.27) is a common enzyme found in wide range of organisms. It catalyzes the interconversion of pyruvate and lactate, with concomitant interconversion of NADH and NAD+. During tissue damage, LDH is released into the bloodstream; therefore it serves as a marker for various diseases and common injuries. BioVision’s PicoProbe™ Lactate Dehydrogenase Activity Assay kit provides a quick and easy method for monitoring Lactate Dehydrogenase activity in a wide variety of samples. In this assay, Lactate Dehydrogenase converts lactate into pyruvate and NADH, which reacts with PicoProbe™ to generate an intense fluorescent product (Ex/Em = 535/587 nm). This kit is simple, highly sensitive and high-throughput adaptable, and can detect Lactate Dehydrogenase activity as low as 1 µU/ml.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty