Would you like to log in again?
β-N-Acetylglucosaminidase Activity Assay Kit (Colorimetric), BioVision Inc.
Supplier: BioVision
AB204705-100TEST
76022-268EA 727.77 USD InStock
76022-268
β-N-Acetylglucosaminidase Activity Assay Kit (Colorimetric), BioVision Inc.
Assays Enzyme Assays
Convenient & sensitive assay to measure NAG activity
  • Convenient & sensitive
  • The assay can detect as low as 50 µU of NAG activity in a variety of samples
β-N-Acetylglucosaminidase (NAG, EC 3.2.1.52) is a lysosomal enzyme that is expressed in various tissues, including kidney, liver and lungs. NAG can cleave N-acetyl-glucosamine, a monosaccharide derivative of glucose. Its concentration in urine is minimal due to its inability to cross the glomerular basal membrane. Increased concentration of NAG in urine indicates renal tubular cell breakdown. Acute Kidney Injury (AKI) is the sudden loss of kidney functions, causing electrolyte imbalance, and retention of urea and other nitrogenous products. NAG has become one of the most studied and used biomarkers for the detection and diagnosis of AKI. BioVision’s NAG Activity Assay Kit provides a simple and sensitive method for monitoring NAG enzymatic activity. In this assay, NAG uses a synthetic p-nitrophenol derivative (R-pNP) as a NAG substrate and releases pNP which can be measured at absorbance (OD 400 nm). The assay can detect as low as 50 µU of NAG activity in a variety of samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty