Would you like to log in again?
Myeloperoxidase (MPO) Peroxidation Activity Assay Kit, BioVision
Supplier: BioVision
AB273334-100TEST
76022-272EA 486.17 USD InStock
76022-272
Myeloperoxidase (MPO) Peroxidation Activity Assay Kit, BioVision
Assays Enzyme Assays
Highly Sensitive Assay, HTS
  • Quick and Easy protocol
  • Includes specific MPO Inhibitor [4-Aminobenzonic Hydrazide] to delineate MPO activity from total peroxidase activity in the sample.
  • Highly Sensitive assay for measuring MPO less than 2 μU per well
Myeloperoxidase (MPO: EC 1.11.2.2) is a peroxidase abundantly expressed in neutrophil granulocytes. It catalyzes the hydrogen peroxide (H2O2) dependent oxidation of chloride anions (Cl-) to generate hypochlorite (HClO). During respiratory burst, neutrophils utilize MPO activity to exhibit cytotoxicity against tumor cells and pathogenic microorganisms. MPO also oxidizes various substances such as phenols and anilines, which can lead to inflammation. Increased MPO levels are associated with an increased risk of cardiovascular disease and myocardial infarction. Thus measurement of MPO peroxidation activity is crucial for disease prediction and therapeutics. BioVision’s Myeloperoxidase (MPO) Peroxidation Activity Assay Kit, oxidizes a substrate to generate fluorescence (Ex/Em = 535/587 nm), which is directly proportional to total peroxidase activity in the sample. A specific MPO inhibitor is provided, which suppresses peroxidase activity due to MPO. This permits differentiation of MPO-mediated peroxidation from the activity of other peroxidases which may be present in the samples. Our kit provides a sensitive, quick and easy method for detecting MPO activity. The assay is high-throughput adaptable and can detect less than 2 µU of MPO activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty