Would you like to log in again?
Purine Nucleoside Phosphorylase Activity Assay Kit (Fluorometric) , BioVision
Supplier: BioVision
AB204706-100TEST
76022-276EA 1350.97 USD InStock
76022-276
Purine Nucleoside Phosphorylase Activity Assay Kit (Fluorometric) , BioVision
Assays Enzyme Assays
The most sensitive kit on the market
  • Rapid, simple & convenient
  • Limit of quantification is 0.005 µU recombinant Purine Nucleoside Phosphorylase
Purine Nucleoside Phosphorylase (PNP) (E.C. 2.4.2.1.) is an enzyme involved in purine metabolism and it catalyzes the cleavage of the glycosidic bond of ribo- or deoxyribonucleosides, in the presence of inorganic phosphate as a second substrate, to generate the purine base and ribose-1-phosphate or deoxyribose-1-phosphate. It is one of the enzymes of the nucleotide salvage pathways that allows the cell to produce nucleotide monophosphates when the de novo synthesis pathway has been interrupted or is non-existent (as is the case in the brain). PNP is a cytosolic enzyme. PNP deficiency disease leads to toxic buildup of deoxyguanosine in T-cells leading to T-lymphocytopenia with no apparent effects on B-lymphocyte function. Inhibition of PNP is an important target for chemotherapeutic applications and treatment of T- cell mediated autoimmune diseases. PNP deficiency is also associated with neurological problems. In BioVision’s Purine Nucleoside Phosphorylase Activity Assay, hypoxanthine formed from the breakdown of inosine is detected via a multi-step reaction, resulting in the generation of an intermediate that reacts with the PNP Probe. The fluorescent product is measured at Ex/Em = 535/587 nm. Limit of quantification is 0.005 µU recombinant Purine Nucleoside Phosphorylase.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty