Would you like to log in again?
PicoProbe™ Hexokinase Activity Assay Kit (Fluorometric), BioVision Inc.
Supplier: BioVision
AB211103-100TEST
76022-280EA 1034.23 USD InStock
76022-280
PicoProbe™ Hexokinase Activity Assay Kit (Fluorometric), BioVision Inc.
Assays Enzyme Assays
Applicatioons are measurement of Hexokinase activity in various tissues/cells, analysis of Glucose metabolism and Cell signaling in various cell types and screening anti-diabetic drugs.
  • Simple and rapid procedure
  • Highly Sensitive
  • Can detect hexokinase activity even as low as 2 µU
Hexokinase (EC 1.1.1.49; also called HK, 6-Phosphate Glucose Kinase, ATP: D-Hexose 6-Phosphotransferase, ATP-dependent Hexokinase) is found in many organisms including bacteria, plants and mammals. Hexokinases play an important role in glucose metabolism. There are four isoforms (HK-I, II, III and IV). HK-I, HK-II, and HK-III have low Km, while HK-IV (also called Glucokinase) has 100-fold high Km for glucose. Hexokinase deficiency leads to diseases such as X-linked muscular dystrophy and rare autosomal recessive hemolytic anemia. On the other hand, increased hexokinase activity is detected in various human tumors and is associated with metastasis. BioVision’s PicoProbe™ Hexokinase Activity Assay kit provides a quick and easy method for monitoring HK activity in a wide variety of samples. In this assay, HK converts glucose into glucose-6-Phosphate, which in turn undergoes a series of reactions and reduce PicoProbe™ to generate intense fluorescent product (Ex/Em = 535/587 nm). The assay is simple, sensitive and high-throughput adaptable and can detect as low as 2 µU of HK activity.
Kit components:
• HK Assay Buffer
• PicoProbe™ (in DMSO)
• HK Substrate
• ATP
• HK Enzyme
• HK Developer
• HK Positive Control
• NADPH Standard (200 nmol)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty