Would you like to log in again?
Rat Renin Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB284512-100TEST
76022-290EA 1363.63 USD InStock
76022-290
Rat Renin Inhibitor Screening Kit (Fluorometric), BioVision
Assays Enzyme Assays
Sensitive Assay to Screen Rat Renin Inhibitors, HTS
  • Rapid & HTS
  • Reliable test for screening/studying/characterizing potential inhibitors of rat renin
  • Includes Inhibitor Control
Renin (EC 3.4.23.15), also known as an angiotensinogenase, is an enzyme that participates in the renin-angiotensin system (RAS) which mediates extracellular volume (i.e. blood plasma, lymph and interstitial fluid), and arterial vasoconstriction. An over-active renin-angiotensin system leads to vasoconstriction and retention of sodium and water, causing hypertension. Renin inhibitors are widely used for the treatment of hypertension. BioVision’s Rat Renin inhibitor screening Kit uses a synthetic peptide substrate with a fluorophore (EDANS) at one end and a quencher (DABCYL) at the other end. Renin catalyzes the cleavage of FRET substrate resulting in a product that is detected fluorometrically at Ex/Em = 328/552 nm. In the presence of a Rat Renin inhibitor, the rate of hydrolysis of the substrate is decreased or inhibited. The kit provides a rapid, simple, sensitive, and reliable test suitable for high-throughput screening of rat renin inhibitors. It is also adaptable to a 384-well format.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty