Would you like to log in again?
Rat Renin Activity Fluorometric Assay Kit, BioVision
Supplier: BioVision
AB284559-100TEST
76022-294EA 1164.45 USD InStock
76022-294
Rat Renin Activity Fluorometric Assay Kit, BioVision
Assays Enzyme Assays
Convenient method to detects Rat Renin Activity.
  • Rapid, simple & convenient
  • The assay can detect rat renin activity as low as 0.12 mU recombinant rat renin.
Renin (EC 3.4.23.15), also known as angiotensinogenase, is an enzyme that participates in the renin-angiotensin system (RAS) which mediates extracellular volume (i.e. blood plasma, lymph and interstitial fluid), and arterial vasoconstriction. In addition to the systemic RAS, locally expressed RAS has been found in the kidney, heart and nervous system. An overactive renin-angiotensin system leads to vasoconstriction and retention of sodium and water, causing hypertension. Renin inhibitors are widely used for the treatment of hypertension. In BioVision’s Rat Renin Activity Assay Kit, Rat Renin and other proteases hydrolyze a FRET substrate resulting in fluorescence at Ex/Em = 328/552 nm. In the presence of a Rat Renin-Specific Inhibitor, hydrolysis of the substrate is due to only the activity of other proteases. The difference between the total activity and the activity in the presence of Rat Renin-Specific Inhibitor gives the Rat Renin Activity in the sample. This rapid, simple & sensitive kit can detect renin activity as low as 0.12 mU recombinant rat renin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty