Would you like to log in again?
Adenosylhomocysteinase (AHCY) Activity Fluorometric Assay Kit, BioVision Inc.
Supplier: BioVision
AB197002-100TEST
76022-296EA 1478.57 USD InStock
76022-296
Adenosylhomocysteinase (AHCY) Activity Fluorometric Assay Kit, BioVision Inc.
Assays Enzyme Assays
Only kit on the market to measure AHCY activity
  • Rapid, simple & convenient
  • The assay can detect AHCY activity as low as 1 µU recombinant human AHCY.
Adenosylhomocysteinase (AHCY) (EC 3.3.1.1) or S-adenosylhomocysteine hydrolase (SAHH); is an enzyme that catalyzes the reversible hydrolysis of S-Adenosyl Homocysteine (SAH) to adenosine and homocysteine. AHCY regulates the intracellular SAH concentration which in turn regulates S-adenosyl methionine (SAM)-dependent methyltransferases. Down-regulation of AHCY has been associated with certain forms of cancer and Huntington’s disease, while in Wilson’s disease; the enzyme is inhibited by the accumulated copper. Mutations in the AHCY gene cause SAHH deficiency disease. BioVision’s AHCY Activity Assay kit can kinetically measure AHCY activity by detecting adenosine generation resulting from the hydrolysis of SAH. Adenosine is detected via a multi-step reaction, resulting in the generation of an intermediate that reacts with the OxiRed™ Probe. The fluorescent product is measured at Ex/Em = 535/587 nm. Limit of quantification (L.O.Q) is 1 µU recombinant human AHCY.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty