Would you like to log in again?
EZCellTM Cell Migration/Chemotaxis Assay Kit (24-well, 3 µm), BioVision
Supplier: BioVision
AB235899-12TEST
76022-308EA 674.18 USD InStock
76022-308
EZCellTM Cell Migration/Chemotaxis Assay Kit (24-well, 3 µm), BioVision
Assays Cellular Assays
Highly Sensitive and Quantitative Assay to measure Cell Migration/Chemotaxis using a Boyden Chamber
  • Highly sensitive fluorometric method to measure cell migration/chemotaxis in response to a variety of biochemical stimuli
  • Simple & High throughput-adaptable
  • Reproducible, Quantitative tool for screening, studying, and characterizing compounds that affect cell migration
Cell migration is a process by which cells move from one location to another. Cell motility is observed in unicellular organisms, and is essential for the development and maintenance of multicellular organisms. Cells often migrate in response to specific external stimuli, including chemical & mechanical signals. Errors during this process have serious consequences, including intellectual disability, vascular disease, tumor formation and metastasis. BioVision’s Cell Migration/Chemotaxis Assay Kit utilizes a Boyden chamber, where the cells migrate through a semipermeable membrane under different stimuli. Cell migration can be analyzed directly by reading fluorescence (Ex/Em = 530/590 nm) in a plate reader. Our assay is easy to use, sensitive and adaptable to high-throughput systems.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty