Would you like to log in again?
EZCellTM Cell Cycle Analysis Kit, BioVision
Supplier: BioVision
AB287852-100TEST
76022-316EA 536.27 USD InStock
76022-316
EZCellTM Cell Cycle Analysis Kit, BioVision
Assays Cellular Assays
Highly Sensitive, HTS compatible, Flowcytometry-based assay
  • Highly sensitive flow cytometry-based method to detect and monitor cells at various stages of the cell cycle
  • Simple & High throughput-adaptable
  • Easy tool for screening compounds that affect cell growth and division
Cell cycle is a ubiquitous, complex sequence of events leading to growth and proliferation of cells. Cell Cycle progression is tightly regulated due to its involvement in development, DNA damage and repair, etc. Anomalies in cell cycle progression can lead to tissue hyperplasia and diseases such as cancer. Cell cycle can be subdivided into interphase (G0/G1, S and G2) and mitotic (M) phase (prophase, metaphase, anaphase and telophase). BioVision’s Cell Cycle Analysis Kit provides a quick and easy method to detect the number of cells in a cell population, which are at a specific stage of the cell cycle. Our kit utilizes a nuclear dye, the binding of which to nucleic acids in the cell results in fluorescence signal, which is proportional to cellular DNA content. The percentages of cells in different phases of the cell cycle (G0/G1, S, and G2/M) can be quantified by flow cytometry. Our method is non-radioactive, rapid and accurate and can be used for high throughput cell cycle analysis with contemporary flow cytometer instruments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty