Would you like to log in again?
TNF alpha (Mouse) ELISA Kit, BioVision
Supplier: BioVision
AB285327-96TEST
76022-318EA 1555.16 USD InStock
76022-318
TNF alpha (Mouse) ELISA Kit, BioVision
Assays ELISAs
Sensitive, Colorimetric Assay
  • Easy, convenient and time-saving method to assay for mouse TNF alpha.
  • Detection Range:15.6 pg/ml – 1000 pg/ml (cell culture supernates), 7.8 pg/ml – 500 pg/ml (mouse serum, plasma). Sensitivity: < 1 pg/ml.
  • This ELISA kit shows no cross-reactivity with other relevant proteins.
BioVision’s mouse TNF alpha ELISA Kit is based on the standard sandwich enzyme-linked immunosorbent assay technology. This assay employs a monoclonal antibody specific for mouse TNF alpha coated on a 96-well plate. Standards and test samples are added to the wells and TNF alpha present in a sample is bound to the wells by the immobilized antibody. A biotinylated detection polyclonal antibody from goat specific for TNF alpha is added subsequently. After washing away the unbound biotinylated antibody with PBS or TBS buffer, Avidin-Biotin-Peroxidase Complex is added to the wells. The wells are again washed with PBS or TBS buffer to remove the unbound conjugates. HRP substrate TMB is used to visualize the HRP enzymatic reaction. TMB is catalyzed by HRP to produce a blue color product that changes into yellow after adding acidic stop solution. The density of yellow color is proportional to the mouse TNF alpha captured onto the plate. This ELISA kit shows no cross-reactivity with other relevant proteins. Detection Range: 15.6 pg/ml – 1000 pg/ml (cell culture supernates), 7.8 pg/ml – 500 pg/ml (mouse serum, plasma). Sensitivity: < 1 pg/ml.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty