Would you like to log in again?
Allure® Silica HPLC Columns, Restek
Supplier: Restek
Allure®
916050210 9160512 9160565 9160572 9160555 916050212 916051E 916057E 9160532 916053E 9160562 916055E 916052E 9160535 916056E 9160552 9160515 9160575 9160525 9160522
10838-252EA 603.17 USD InStock
10838-252 10839-408 10839-406 10839-418 10839-426 10840-624 10838-268 10839-424 10840-616 10839-444 10839-442 10839-420 10839-432 10839-430 10837-568 10837-566 10840-632 10840-210 10838-260 10840-212
Allure® Silica HPLC Columns, Restek
Chromatography Columns
Allure® Silica HPLC columns contain a highly retentive phase for normal phase separations.
  • Very high surface area
  • Type B silica packing
Similar phases: Maxsil Si (irregularly shaped particle), LiChrospher Si-60 (irregularly shaped particle).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty