Would you like to log in again?
ImmunO™ Rat Serum, MP Biomedicals
Supplier: MP Biomedicals
ImmunO™
08642941
IC642941EA 131.73 USD InStock
IC642941
ImmunO™ Rat Serum, MP Biomedicals
Sera (Serum)
Normal rat serum may be used as a blocking agent and controls in immunoassays
  • Sterile filtered
When reconstituted with 10 mL (sterile) distilled water or buffer, as appropriate for application, protein concentration will be at 5.5 - 7.0 gm/dL.
This rat serum is pooled from a normal donor population.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty