Would you like to log in again?
WesternSure® Pre-stained Chemiluminescent Protein Ladder, LI-COR®
Supplier: LI-COR®
926-98000
102971-216EA 276.92 USD InStock
102971-216
WesternSure® Pre-stained Chemiluminescent Protein Ladder, LI-COR®
Electrophoresis Reagents Molecular Weight Markers
WesternSure® chemiluminescent protein ladder is the only pre-stained, multi-colored protein ladder that provides film and digital detection.
  • Fits with your current protocol without optimization
  • Does not rely on strep-tagged HRP or secondary antibody binding
  • Works with a variety of substrates
  • Provides a vibrant colour scheme that allows you to quickly orient your membrane
Ladder consists of 9 recombinant protein bands (8-260 kDa); 1 green reference band (8 kDa); 3 orange reference bands (38, 70, 260 kDa).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty