Would you like to log in again?
Masterflex® L/S C-Flex Tubing, Cole Parmer
Supplier: Cole Parmer
06424-17 06424-16 06424-82 06424-35 06424-25 06424-24 06424-36 06424-13 06424-14 06424-18 06424-73 06424-15 06424-26
76049-390PK 206.56 USD InStock
76049-390 76049-382 76049-394 76049-380 76049-392 76049-386 76049-398 76049-376 76049-384 76049-374 76049-396 76049-388 76049-378
Masterflex® L/S C-Flex Tubing, Cole Parmer
Tubing Peristaltic Pump Tubing
Combines the biocompatabilty of silicone with the chemical resistance similar to PVC.
  • Very low protein binding
  • Heat sealable and weldable
  • Exceptional tensile and tear strength
  • Good biocompatibility
  • Low gas permeability
  • Smooth surface
  • Opaque white finish
Masterflex® L/S pump tubing is manufactured to extremely close tolerances that match the L/S pump heads, ensuring accurate, repeatable flow and long tubing life.
Applications include general-purpose laboratory use requiring high performance and long life.
Operating temperture range is –40 to -60 °C. Static temperature range is –60 to 121 °C.
Sterilize by EtO, autoclave, or gamma radiation. When autoclaving, do not clamp; wrap in non-linting cloth or paper at MAX 132 °C- 1 bar for 30 minutes.
Certifications: USP Class VI; European Pharmacopeia (3.2.9); FDA; Exceeds 3A Sanitary; Reach Compliant, RoHS Compliant, ADCF Compliant.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty