Would you like to log in again?
HistoMark® TrueBlue™ Peroxidase Substrate Kit, LGC Clinical Diaganostics, Inc
Supplier: LGC Clinical Diagnostics
5510-0035
95059-336EA 482.17 USD InStock
95059-336
HistoMark® TrueBlue™ Peroxidase Substrate Kit, LGC Clinical Diaganostics, Inc
Assays Immunological Assays
This kit provides the most sensitive detection for peroxidase systems utilizing TrueBlue Substrate
TrueBlue forms a blue chromogenic product, providing excellent contrast with DAB and other substrates for multiple labeling experiments.
TrueBlue is 50-100 times more sensitive than DAB and is non-carcinogenic. Strongly recommended for the detection of least-abundant markers.
Delivery information: Kit includes TrueBlue Substrate Solution, Orcein Counterstain, and Blocking Solution Concentrate for blocking endogenous peroxidase activity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty