Would you like to log in again?
Tryptic Soy Broth Dehydrated Culture Media, BD Diagnostics
Supplier: BD Diagnostics
257488 257488
95062-836PK 207.23 USD InStock
CA95062-836 95062-836
Tryptic Soy Broth Dehydrated Culture Media, BD Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Tryptic Soy Broth are nutritious media that will support the growth of a wide variety of microorganisms, including common aerobic and facultatively anaerobic bacteria and fungi.
It is used in qualitative procedures for the detection, isolation, and cultivation of fastidious and nonfastidious microorganisms, in sterility testing, and in the microbiological examination of nonsterile products.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty