Would you like to log in again?
SP Wilmad-LabGlass Squibb Separatory Funnels, SP Industries
Supplier: SP Industries
LG-8435T-106 LG-8435T-104 LG-8435T-102
14203-336EA 359.11 USD InStock
14203-336 14207-406 14210-584
SP Wilmad-LabGlass Squibb Separatory Funnels, SP Industries
Funnels Separating Funnels
These economy style squibb separatory funnels come complete with a detachable PTFE stopcock, glass drip tip, and plastic stopper.
Ordering information: Glass tip (LG-3958-106) and PTFE valve (3958T-104) must be purchased together for proper fit.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty