Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyacrylonitrile, M.W. 150000

Supplier: Pfaltz & Bauer

P21470-100G
101179-380EA 1005.74 USD InStock
101179-380
Polyacrylonitrile, M.W. 150000
Polyacrylonitrile


Formula: (C₃H₃N)n
Boiling Pt: 77…78 °C
Melting Pt: 317 °C
Density: 1,184 g/cm³
MDL Number: MFCD00084395
CAS Number: 25014-41-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty