Would you like to log in again?
Keto-Diastix® Reagent Strips for Urinalysis, Ascensia Diabetes Care
Supplier: Ascensia Diabetes Care (BAYER)
Keto-Diastix®
2883
AM2883CS 676.36 USD InStock
AM2883
Keto-Diastix® Reagent Strips for Urinalysis, Ascensia Diabetes Care
Clinical Diagnostic Supplies Clinical Reagents and Kits
Keto-Diastix® is a ready-to-use strip with two reagent areas specifically designed to provide a fast, convenient way to test urine for glucose and ketone.
  • Easy dip testing for urine glucose
  • Accurate timing is essential to provide reliable results
When the strip is dipped into a fresh urine sample one area of the strip will change color if it detects glucose. The other area of the strip will change color if it detects ketones. The colors will vary depending on the amount of each substance found in the urine.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty