Would you like to log in again?
MangoTaq™ DNA Polymerase, Bioline
Supplier: Bioline
MangoTaq™
BIO-21083
490015-790EA 468.44 USD InStock
490015-790
MangoTaq™ DNA Polymerase, Bioline
Nucleic Acid Reagents End-point PCR Enzymes and Kits
MangoTaq™ DNA Polymerase is a formulation of Taq DNA Polymerase that offers high-yield across a wide range of DNA concentrations.
  • Direct gel loading - no need for further post-PCR processing steps
  • Easy visual recognition- reduces pipetting errors
  • Robust performance - perfect for a wide range of PCR reactions
  • Reproducible results - consistent QC ensures reliability
It possesses 5' to 3' exonuclease activity and leaves an 'A' overhang, resulting in PCR product suitable for effective integration into TA cloning vectors
The colored reaction buffer contains red and orange dyes, which separate during electrophoresis and provide quick reference points for monitoring the mobility of the DNA samples in the gel. The colored reaction buffer can be loaded directly onto an agarose gel for analysis without the need for separate gel-loading buffer. The presence of the dyes has no effect on routine enzymatic manipulations, although extremely rare exceptions may exist.
Since the colorless reaction buffer does not contain reference dyes, it is suitable for use when reaction products will be used directly for down-stream processes involving absorbance or fluorescent detection. The specificity and performance of MangoTaq DNA Polymerase can be further improved with the use of 3% DMSO, which is designed for GC or AT- rich DNA, "dirty" templates or sequences with a high level of secondary structure.
The polymerase is supplied with two different reaction buffers for greater flexibility. For high-throughput applications, MangoTaq and the colored reaction buffer make an ideal choice, since this combination enables the user to load directly on a gel in order to facilitate easy recognition.
Delivery information: Includes 5X colored reaction buffer(Red and Orange dyes), 5X colorless reaction buffer(No reference dyes).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty