Would you like to log in again?
Magid® Bib Apron, Magid Glove
Supplier: Magid
1859
69100-069EA 15.39 USD InStock
69100-069
Magid® Bib Apron, Magid Glove
Workwear
Flame resistant bib aprons are made from fabric self extinguishes when the ignition source or flame has been removed.
  • SparkGuard® flame resistance
  • Made from 100% cotton sateen
This green bib apron has 6 snaps that firmly attach on to Magid® cape sleeve for extended protection.
Certifications: ASTM D6413.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty