Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Antibody Diluent / Block, 1X, 100 ml, PerkinElmer

Supplier: Perkin Elmer
ARD1001EA
MSPP-ARD1001EAEA 338.46 USD InStock
MSPP-ARD1001EA
Antibody Diluent / Block, 1X, 100 ml, PerkinElmer
Buffers Assay Buffers
This ready-to-use antibody diluent and blocking buffer formulation has been optimized specifically for Opal™ and TSA® Plus immunohistochemistry protocols. It acts to minimize non-specific interaction of antibodies that can be a source of background.

  • Storage Temperature Description-+2 -> +8 C
  • H phrases US-H317
  • P phrases US-P261-P280g-P272-P363-P333+P313-P302+P352a-P321-P501a
  • GHS-Signal words - US-Warning
  • GHS-Symbols - US-GHS07

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty