Would you like to log in again?
Media Bottles, Anerobic, Clear, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
CLS-4217-03 CLS-4217-05 CLS-4217-04 CLS-4217-01 CLS-4217-02
10170-768EA 132.03 USD InStock
10170-768 10170-766 10170-774 10170-772 10170-770
Media Bottles, Anerobic, Clear, Chemglass
Bottles Cell Culture Bottles
Anaerobic media bottles are designed for maintaining anaerobic conditions, useful for studies involving methane-producing bacteria, cell culture, drug discovery, and research and development.
  • Autoclavable
  • Manufactured from Duran® Schott borosilicate glass
Each bottle has a graduation scale indicating the approximate volumes and has a writing area for identification. Bottle is supplied with a rubber stopper and aluminum seal.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty