Would you like to log in again?
BD Taxo™ SPS Discs, BD Diagnostic
Supplier: BD Diagnostics
BD Taxo™
231726 231726
90002-548PK 153.8 USD InStock
90002-548 CA-90002-548
BD Taxo™ SPS Discs, BD Diagnostic
Microbiology Test Systems Microorganism Tests
BD Taxo™ Differentiation Discs SPS (sodium polyanethol sulfonate) are used for presumptively identifying Peptostreptococcus anaerobius and Gardnerella vaginalis.
These are paper discs that contain sodium polyanethol sulfonate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty