Would you like to log in again?
PassPort® Sterilizer Monitoring Service Kit, Crosstex
Supplier: Crosstex
PS-012 PS-052
101101-778PK 166.92 USD InStock
101101-778 101101-780
PassPort® Sterilizer Monitoring Service Kit, Crosstex
Sterilization Indicators
PassPort® Sterilizer monitoring system provides third party legal documentation of sterilization from the largest sterilizer testing laboratory.
  • EO Gas test results are completed and reported back to the customer after 7 days incubation
  • Steam test results are documented and reported back to the customer after 24 hours incubation
  • Chemical vapor test results are documented and reported back to the customer after 72 hours incubation
  • Allows access to spore test results on-line (www.checkyourtest.com) and includes prepaid envelopes for each test
Two test strips and one control strip are provided for users to spore test multiple locations within the sterilizer chamber.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty