Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Mannosamine hydrochloride

Supplier: TORONTO RESEARCH CHEMICALS MS
Synonyms: (2S,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride

M16700010G M1670001G
100573-508EA 1676.74 USD InStock
100573-508 100573-506
D-Mannosamine hydrochloride
D-Mannosamine hydrochloride


Formula: C₆H₁₄ClNO₅
MW: 215.63 g/mol
Melting Pt: >160 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00064557
CAS Number: 5505-63-5
Order Now
Specification Test Results
Appearance White to Off-White Solid

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty