Would you like to log in again?
FirmWIPE® Cleanroom Sponge Wipes, Foamtec International
Supplier: Foamtec
HT4331
10810-808CS 1502.3 USD InStock
10810-808
FirmWIPE® Cleanroom Sponge Wipes, Foamtec International
Wipes Cleanroom Wipes Dry Cleanroom Wipes
FirmWIPE® is constructed from an open-cell ester-based polyurethane foam which has undergone a special felting process.
  • Open-cell polyurethane structure allows for fiber-free wiping
  • Excellent pick-up of Thick Film inks and solder paste
  • Open-cell, felted foam structure allows for controlled transfer of solvent to optimize cleaning of Thick Film screens
  • Ester-based chemistry and felted construction enable broad solvent compatibility
  • Uniform porosity allows for excellent contamination pickup
FirmWIPE® wipes provides fiber-free wiping in applications where durability and fast wicking, without dripping, are desirable. Due to its ability to conform to metal mask surfaces, wipe enables excess paste to be thoroughly removed while minimizing abrasion to the emulsion. Its unique pore structure makes it ideal for cleaning metal masks used to print LTCC components, capacitors, EL, OLED, and Plasma Display Panels.
Certifications: Cleanroom Class 100 configurations.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty