Would you like to log in again?
3M™ Industrial Citrus Base Cleaner, 3M
Supplier: ORS Nasco
3M™
5011132181
300005-773CS 730.37 USD InStock
300005-773
3M™ Industrial Citrus Base Cleaner, 3M
Cleaning Agents Detergents and Surface Cleansers
A multi-purpose cleaner which will dissolve and flush away dirt, grease, tar, most non-curing type adhesive films, and similar substances.
  • Citrus oil based cleaner that emulsifies when flushed with water
  • Contains no petroleum distillates
  • Contains no methylene chloride, 1, 1, 1-trichloroethane or chlorofluorocarbons
Use on metal parts, tools, motors, and concrete. 91% VOC. Fresh citrus scent.
Removes grease, oil, ink, and most adhesives.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty