Would you like to log in again?
Disposable Mop Heads C1, Contec®
Supplier: Contec
C1-MOP
68310-378CS 404.95 USD InStock
68310-378
Disposable Mop Heads C1, Contec®
Mops
Made from non woven polyester and cellulose fibers, these mop heads absorb liquids quickly leaving mopped surfaces almost dry.
  • Sterilizable by autoclave, ethylene oxide, gamma radiation methods
  • For use with Fiberglass Handle (68310-048)
Packaging: Double bagged.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty