Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Magnetic Bar Magnets
Supplier: United Scientific Supplies
MPC080
470229-308PR 8 USD InStock
470229-308
Magnetic Bar Magnets
Magnets
Plastic coated bar magnets.
  • Color-coded ends
  • Strong Alnico magnets
These alnico magnets are encased in red and blue plastic to provide extra durability. Each magnet measures 80x15x10mm. Shipped as a pair.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty