Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vitamin B12 (cyanocobalamin)

Supplier: MORAVEK BIOCHEMICALS MS

MT1953-50UCI
101972-854EA 1516.92 USD InStock
101972-854
Vitamin B12 (cyanocobalamin)
Vitamin B12 (cyanocobalamin)


Formula: C₆₃H₈₈CoN₁₄O₁₄P
MW: 1355.38 g/mol
MDL Number: MFCD00151092
CAS Number: 68-19-9
UN: 2910
ADR: ,
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty