Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Midostaurin

Supplier: MORAVEK BIOCHEMICALS MS
Synonyms: PKC412

MT1954-50UCI
101972-856EA 1516.92 USD InStock
101972-856
Midostaurin
Midostaurin


Formula: C₃₅H₃₀N₄O₄
MW: 570.649 g/mol
MDL Number: MFCD12828879
CAS Number: 120685-11-2
UN: 2910
ADR: ,
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty