Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-Deoxyadenosine, [2,8-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT641-1MCI MT641-250UCI
101973-272EA 3125.38 USD InStock
101973-272 101973-270
2'-Deoxyadenosine, [2,8-3H] ≥97% (by HPLC), MORpure™
2'-Deoxyadenosine, [2,8-3H]


CAS Number: 131211-89-7
UN: 2910
ADR: ,
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty