Would you like to log in again?
Biologix Serological Pipettes
Supplier: TradeWinds (TWD)
07-5005 BX07-5050 07-5010
76081-576CS 441.04 USD InStock
76081-576 76081-572 76081-570
Biologix Serological Pipettes
Pipettes Serological Pipettes
Color coded polystyrene serological pippetes come in sizes ranging from 1 to 50 ml.
  • Highly transparent polystyrene
  • Six volumes available
  • Individually wrapped packaging
  • Sterile
  • Ascending and descending graduations
These color-coded serological pipettes have ascending and descending graduations.
Certifications: ISO Certified: GB/T 19001-2008/ISO 9001:2008.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty