Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Congo red

Synonyms: Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulfonate)Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate)Direct Red 28Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulfonate)Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate)CI 22120

AB145645-500MG AB145645-100MG
76021-142EA 122.58 USD InStock
76021-142 76021-144
Congo red
Congo Red
Congo Red amyloid fibril-binding dye; inhibits Fib-beta A neurotoxicity by inhibiting fibril formation or by binding to preformed fibrils

Congo red also inhibits the pancreatic islet cell toxicity of diabetes-associated amylin, another type of amyloid fibril. Also acts as an inhibitor of protease-resistant prion protein formation.


Formula: C₃₂H₂₂N₆Na₂O₆S₂
MW: 696.67 g/mol
MDL Number: MFCD00004028
CAS Number: 573-58-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/21/13/23492113.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty