Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Exonuclease VII
Supplier: Lucigen
EN510250
76085-630EA 473.77 USD InStock
76085-630
Exonuclease VII
Enzymes
ssDNA exonuclease with both 5´ to 3´ and 3´ to 5´ exonuclease activities
  • Remove single-stranded oligonucleotide primers after PCR
  • Minimize the effect of primers left over from previous PCR amplifications
  • Heat inactivate the enzyme at 95°C for 10 minutes
Exonuclease VII has high enzymatic specificity for ssDNA and exhibits both 5´ to 3´ and 3´ to 5´ exonuclease activities. It is useful for rapid removal of single-stranded oligonucleotide primers from a completed PCR when different primers are required for subsequent PCR amplifications. Exonuclease VII digestion of ssDNA occurs in the absence of magnesium. Exonuclease VII can be inactivated by heating at 95°C for 10 minutes.
Certifications: For research use only. Not for human or diagnostic use.
Ordering information: Contents include enzyme and a 5X reaction buffer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty