Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
5´-Phosphate-Dependent Exonuclease, Lucigen
Supplier: Lucigen
TER51020
76081-784EA 494.11 USD InStock
76081-784
5´-Phosphate-Dependent Exonuclease, Lucigen
Enzymes
A processive 5´ to 3´ riboexonuclease that specifically digests RNA with 5´-monophosphate ends
  • Specific: Processively digests RNA with 5´-monophosphate ends but not RNAs with 5´-triphosphate, 5´-cap or 5´-hydroxyl groups starting from the 5´ end
  • Flexible: Use this enzyme to help characterize 5´-ends of RNA and to enrich for mRNA in prokaryotic or eukaryotic total RNA preparations
Terminator™ 5´-Phosphate-Dependent Exonuclease is a processive 5´ to 3´ exonuclease that digests RNA that has a 5´-monophosphate end
Applications include:
Characterize the 5´ termini of RNA transcripts
Enrich for mRNA in prokaryotic or eukaryotic total RNA preparations
Note: Terminator™ 5´-Phosphate-Dependent Exonuclease has not been optimized for RNA-Seq applications.
It does not digest RNA that has a 5´-triphosphate, 5´-cap or 5´-hydroxyl group. Terminator Exonuclease is not inhibited by proteinaceous RNase inhibitors, such as RiboGuard™ RNase Inhibitor or placental ribonuclease inhibitors.
Certifications: For research use only. Not for human or diagnostic use.
Ordering information: Contents include enzyme and 10X Terminator™ Reaction Buffers A and B.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty