Would you like to log in again?
Premixed dNTP Solutions, PCR-ready dNTP Mix, Biosearch Technologies
Supplier: Biosearch Technologies
D59104 D08104
76081-614EA 184.55 USD InStock
76081-614 76081-616
Premixed dNTP Solutions, PCR-ready dNTP Mix, Biosearch Technologies
Nucleic Acid Reagents Nucleotides
Ready-to-use premixed dNTP solutions for PCR and other DNA synthesis reactions
  • Sterile, neutral pH solutions of premixed dGTP, dATP, dTTP and dCTP
  • Available at two different concentrations of each dNTP (2.5 mM or 25 mM)
Sterile, neutral pH, premixed solutions of dATP, dCTP, dGTP, and dTTP are available for use in DNA synthesis reactions including PCR. The premixed dNTPs solutions are available at two concentrations, either 2.5 mM or 25 mM of each dNTP. Each solution is sufficient for at least 1,000 50-µL reactions at concentrations used in typical reactions with most DNA polymerases.
Certifications: For research use only. Not for human or diagnostic use.
Ordering information: Each solutions contains dATP, dCTP, dGTP and dTTP at the indicated concentrations and volumes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty