Would you like to log in again?
Acetaldehyde Assay Kit , BioAssay Systems
Supplier: BioAssay Systems
EFAC-100
76121-364EA 456.89 USD InStock
76121-364
Acetaldehyde Assay Kit , BioAssay Systems
Assays Cellular Assays
Acetaldehyde determination in biological and food/beverage samples.
  • Fast and sensitive. Linear detection range (50µL sample): 0.5 µM to 60 µM acetaldehyde in 96-well plate assay.
  • The procedure involves adding a single working reagent, and reading the absorbance after 30 minutes.
  • Room temperature assay. No 37°C heater is needed.
  • Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day.
  • Method: FL530/585nm
  • Samples: Biological samples (e.g. plasma, serum, urine, tissue and culture media) and food/beverage samples (e.g. wine, coffee, and juice).
ACETALDEHYDE (CH3CHO) is one of the most widely occurring aldehydes in nature and is commonly used in industry. Acetaldehyde, a metabolic byproduct of ethanol in the liver, is toxic to the human body and is rapidly converted to the less harmful acetic acid by the enzyme aldehyde dehydrogenase. People with a deficiency of aldehyde dehydrogenase accumulate acetaldehyde when consuming alcohol and this accumulation results in facial and body flushing often referred to as “Asian flush syndrome.” Build up of acetaldehyde has also been associated with the effects of hangovers from alcohol consumption. Although classified as a carcinogen, acetaldehyde is naturally found in many foods and beverages such as ripe fruit, coffee, and wine. BioAssay Systems' colorimetric acetaldehyde assay kit is based on aldehyde dehydrogenase catalyzed oxidation of acetaldehyde, in which the formed NADH reduces a formazan reagent. The intensity of the product color, measured at 565 nm, is directly proportional to the acetaldehyde concentration in the sample.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty