Would you like to log in again?
EZBlock™ Protease Inhibitor Cocktail V, EDTA-Free, BioVision
Supplier: BioVision
AB287909-1VIAL AB287909-5VIAL
76022-096EA 352.39 USD InStock
76022-096 76022-094
EZBlock™ Protease Inhibitor Cocktail V, EDTA-Free, BioVision
Cell Culture Supplements and Additives
A cocktail of four protease inhibitors that have been optimized for the inhibition of serine and cysteine but not metalloproteases
Since this cocktail does not contain any EDTA, metal-dependent proteins and IMAC (immobilized metal affinity chromatography) isolation techniques (e.g. for poly-His-tagged proteins) are not affected. Reconstitute each vial with 1 ml D.I water to obtain a 100X concentrated stock solution. A 1X stock solution contains 500 µM of AEBSF, HCl (Cat. No. 1644), 150 nM of Aprotinin, bovine lung (Cat. No. 4690), 1 µM of E-64 (Cat. No. 1739), and 1 µM of Leupeptin, hemisulfate (Cat. No. 1648). Provided with a directional insert.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty