Would you like to log in again?
EZBlock™ Protease Inhibitor Cocktail X, EDTA-Free, BioVision
Supplier: BioVision
AB287910-5VIAL AB287910-1VIAL
76022-124EA 367.73 USD InStock
76022-124 76022-122
EZBlock™ Protease Inhibitor Cocktail X, EDTA-Free, BioVision
Cell Culture Supplements and Additives
A cocktail of six protease inhibitors with broad specificity for the inhibition of serine, cysteine, and aminopeptidases
Recommended for general use. (Note: One vial is recommended for the inhibition of proteases present in 20 g of cell extract). Reconstitute each vial in 100 ml D.I. water to obtain 100 ml of the cocktail solution. The reconstituted cocktail solution contains 2 mM of AEBSF, HCl (Cat. No. 1644), 0.3 µM of Aprotinin, bovine lung (Cat. No. 4690), 14 µM of E-64 (Cat. No. 1739), 1 µM of Leupeptin, hemisulfate (Cat. No. 1648), and 130 µM of Bestatin (Cat. No. 1733). Provided with a directional insert.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty